Learn More
Bachem Fmoc-p-azido-Phe-OH
SDP
Supplier:  Bachem 4020251.0001
Encompass_Preferred
Fmoc-p-azido-Phe-OH, 163217-43-4, C24H20N4O4, Mr 428.45, 1g. The azido group is sensitive towards piperidine. In Fmoc-SPPS, this derivative is best introduced as N-terminus or modified before elongating the peptide. Fmoc-p-azidophenylalanine may be inserted as a precursor of p-aminophenylalanine. Levinson et al. reduced the azido group on-resin with a large excess of tributylphosphine in DMF.
Catalog No. 50-194-7258