Learn More
STEMCELL Technologies "The Big Easy" EasySep™ Magnet, For isolating: 1 Unit
SDP
Catalog No. NC0056459
Encompass
"The Big Easy" EasySep™ Magnet is designed for cell separation procedures using EasySep™ reagents when processing a large number of cells, or when using RoboSep™. "The Big Easy" EasySep™ Magnet generates a high-gradient magnetic field in the interior cavity that is strong enough to separate cells labeled with EasySep™ Magnetic Particles without the use of columns. "The Big Easy" EasySep™ Magnet is designed to hold a standard 14 mL (17 x 95 mm) polystyrene tube.
Catalog No. NC0056459 Supplier STEMCELL Technologies Supplier No. 18001
"The Big Easy" EasySep™ Magnet is designed for cell separation procedures using EasySep™ reagents when processing a large number of cells, or when using RoboSep™. "The Big Easy" EasySep™ Magnet generates a high-gradient magnetic field in the interior cavity that is strong enough to separate cells labeled with EasySep™ Magnetic Particles without the use of columns. "The Big Easy" EasySep™ Magnet is designed to hold a standard 14 mL (17 x 95 mm) polystyrene tube.