Learn More
INNOSYNTH TECHNOLOGIES LLC BioHeight™ 5x5” Polycarbonate 4-in-1 Adaptive-Height Cryo Box
Catalog No. NC3856723
Encompass_Preferred
The BioHeight™ control-layer leverages VHT™ to standardize storage across dimensions. This 36-cell infrastructure replaces 3-5 legacy SKUs (25, 81, or 100-place), increasing capacity from 65% to 95%.
Catalog No. NC3856723 Supplier INNOSYNTH TECHNOLOGIES LLC Supplier No. BH5PC
The BioHeight™ control-layer leverages VHT™ to standardize storage across dimensions. This 36-cell infrastructure replaces 3-5 legacy SKUs (25, 81, or 100-place), increasing capacity from 65% to 95%.