Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lux Blox™ The Build Anything Bin
SCI_ED

Catalog No. S81701
Change view
Click to view available options
Grade:
Pre-K to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S81701 Pre-K to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S81701 Supplier Lux Blox™ Supplier No. LUXSTB
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Kit contains 1000 pieces allowing kids to create virtually anything! Provides children a creative outlet to help them learn and retain the perfect balance of physical, emotional, and cognitive stimulation.

  • Create a guitar, spaceship, castle, even a near life-sized boat!
  • Connections are made through linking which provides kids the immediate experience in the world of kinematics
  • Includes bags upon bags full of brilliant LUX colors, all in secured in a durable storage tote
  • LUX is a Parents' Choice Gold Award Winner, named Top Toy in 2018 and 2019 by Autism Live
  • Used by many educators for teaching STEAM in classrooms (Science, Technology, Engineering, Art and Math)
  • ABS Plastic is tested safe and presents no choking hazard

Specifications

Color Multi
Grade Pre-K to 12
Age 6
Includes 100 Ea. of Neon Orange, Teal Blue, Neon Green, Purple, Royal Blue, Neon Pink, Yellow, Red, White and Black in a Durable Storage Tote
Material ABS Plastic
Quantity 1 Kit
Content And Storage Durable Storage Bin
Product Type Build Anything Bin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.