Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science The Carbon Cycle in Soil
GREENER_CHOICESCI_ED

Catalog No. S139201 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S139201 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139201 Supplier Innovating Science Supplier No. IS9759
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

In this activity, students will learn about the carbon cycle in soil and how soil impacts global warming.

  • Changes in the carbon cycle and climate due to human activities have had detrimental effects on the environment. Through 3 different activities, students will investigate one cause of global warming, and the effects of a positive feedback loop.
  • Kit contains enough materials for 15 groups of students

Specifications

Grade 5+
Includes Instructional manual and all necessary chemicals
Class Size 15 Lab Groups
Product Type Educational Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.