Learn More
Bachem Octaarginine Trifluoroacetate
SDP
Supplier:  Bachem 4065816.0001
Encompass_Preferred
Octaarginine Trifluoroacetate, H-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-OH, 148796-86-5, C48H98N32O9, Mr 1267.52, 1mg. Octa-Arg. The cell-penetrating peptide octa-arginine is a potent inhibitor of proteasome activities.
Catalog No. 50-194-7446