Learn More
Bachem Collagen Type II Fragment Trifluoroacetate
SDP
Supplier:  Bachem 4030511.1000
Encompass_Preferred
Collagen Type II Fragment Trifluoroacetate, H-Pro-Thr-Gly-Pro-Leu-Gly-Pro-Lys-Gly-Gln-Thr-Gly-Glu-Leu-Gly-Ala-Hyp-Gly-Asn-Lys-Gly-Glu-Gln-Gly-Pro-Lys-OH, 144703-90-2, C106H174N32O37, Mr 2488.74, 1mg. (Ala260,Hyp261,Asn263)-Collagen Type II (245-270). The collagen II fragment PTGPLGPKGQTGELGA-Hyp-GNKGEQGPK, a peptide analog of an antigenic determinant of type II collagen, prevents the onset of collagen-induced arthritis in vivo.
Catalog No. 50-194-7439