Learn More
Brookfield Engineer Labs The Computrac® MAX® 5000XL moisture, solids, and ash analyzer
Catalog No. NC3888308
Encompass
The Computrac® MAX® 5000XL moisture, solids, and ash analyzer can provide accurate readings from a single sample, offering many of the same features of thermogravimetric analyzers at a fraction of the cost. It has the versatility to a wide range of material testing tasks.- Rugged Construction- High Temperature Testing- Multistage Testing- Self-Cleaning Oven- Able to test material up to 100 times faster
Catalog No. NC3888308 Supplier Brookfield Engineer Labs Supplier No. MAX5000XL
Add to Cart
Add to Cart
Request a Quote
The Computrac® MAX® 5000XL moisture, solids, and ash analyzer can provide accurate readings from a single sample, offering many of the same features of thermogravimetric analyzers at a fraction of the cost. It has the versatility to a wide range of material testing tasks.- Rugged Construction- High Temperature Testing- Multistage Testing- Self-Cleaning Oven- Able to test material up to 100 times faster