Learn More
Bachem Fmoc-Glu(OtBu)-OPfp
SDP
Catalog No. 501947172
Encompass_Preferred
Fmoc-Glu(OtBu)-OPfp, 86061-04-3, C30H26F5NO6, Mr 591.53, 5g.
Catalog No. 50-194-7172 Supplier Bachem Supplier No. 4011874.0005
Fmoc-Glu(OtBu)-OPfp, 86061-04-3, C30H26F5NO6, Mr 591.53, 5g.