Learn More
Perkin Elmer US LLC DSC 7 Dry Box Replacement Iris - Qty. 8
Catalog No. 502016351
Encompass_Preferred
The DSC 7 Dry Box Replacement Iris Kit provides eight coverings made of a natural tan-colored gum rubber material with iris-like openings. These coverings are used to replace the worn coverings on the entry points of the DSC 7 Dry Box. Qty. 8
Catalog No. 50-201-6351 Supplier Perkin Elmer US LLC Supplier No. N5191009
The DSC 7 Dry Box Replacement Iris Kit provides eight coverings made of a natural tan-colored gum rubber material with iris-like openings. These coverings are used to replace the worn coverings on the entry points of the DSC 7 Dry Box. Qty. 8