Learn More
Bachem (Gln22)-Amyloid Beta Protein (1-42) Trifluoroacetate
SDP
Supplier:  Bachem 4050945.0500
Encompass_Preferred
(Gln22)-Amyloid Beta Protein (1-42) Trifluoroacetate, H-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Gln-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-Ile-Ala-OH, 147335-12-4, C203H312N56O59S, Mr 4513.12, 0.5mg. Amyloid b-Protein (1-42) Dutch Mutation, Amyloid b-Protein (1-42) E22Q, Amyloid-beta Peptide(1-42), E22Q, (human), Amyloid-beta Protein (1-42) (human), Dutch Mutation, [Gln22] - beta - Amyloid (1 - 42), [Gln22]-Abeta (1-42), (human). The Dutch mutation (E22Q) aggregates more readily than the wild-type sequence. The resulting fibrils show increased neurotoxicity.
Catalog No. 50-194-6610