Learn More
Izon Science Usa Ltd Izon Exoid Nanoparticle Measurement Device, Tunable Resistive Pulse Sensing, Single Particle Measurement, Semi-Automated, 10.8 kg, ISO 13485:2016 Cert, 40 nm to 11 μm Size Range, 1E5 to 1E11 particles/mL
SDP
Catalog No. NC2092286
Encompass
The Izon Exoid (EX1) is an innovative nanoparticle measurement device designed for superior characterization of complex biological and nanomedicine samples. It utilizes advanced Tunable Resistive Pulse Sensing (TRPS) technology to offer high-resolution single-particle measurements, making it a significant upgrade over traditional methods. The semi-automated device streamlines workflows, improving usability and sensitivity for a variety of nanoparticle types. It is certified to ISO 13485:2016 standards, ensuring quality and reliability in data collection for research applications.
  • Technology based on Tunable Resistive Pulse Sensing (TRPS)
  • Capable of measuring particle size, concentration, and zeta potential
  • Resolution focused on single particle data
  • Size range of 40 nm to 11 μm
  • Concentration range from 1E5 to 1E11 particles/mL
  • Compatible with externa...
This item is not returnable. View return policy
Catalog No. NC2092286 Supplier Izon Science Usa Ltd Supplier No. EX1
Add to Cart
Add to Cart
Request a Quote
The Izon Exoid (EX1) is an innovative nanoparticle measurement device designed for superior characterization of complex biological and nanomedicine samples. It utilizes advanced Tunable Resistive Pulse Sensing (TRPS) technology to offer high-resolution single-particle measurements, making it a significant upgrade over traditional methods. The semi-automated device streamlines workflows, improving usability and sensitivity for a variety of nanoparticle types. It is certified to ISO 13485:2016 standards, ensuring quality and reliability in data collection for research applications.
  • Technology based on Tunable Resistive Pulse Sensing (TRPS)
  • Capable of measuring particle size, concentration, and zeta potential
  • Resolution focused on single particle data
  • Size range of 40 nm to 11 μm
  • Concentration range from 1E5 to 1E11 particles/mL
  • Compatible with externa...