Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ The Field Explorer Kit
GREENER_CHOICESCI_ED

Catalog No. S89528 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S89528 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S89528 Supplier Supertek™ Supplier No. PH96029S3
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inspiring Kit for the budding outdoor scientists! The Field Explorer Kit sends you on your way for a perfect field trip by offering all the companion components you will need, including a journal to record all that you find!

  • Perfect field trip companion
  • Backpack filled with:
    • 1-20X Stereo Microscope, battery operated
    • 1-Large Insect Net with telescoping handle.
    • 1-Aqua Net
    • 1-Bug Viewer with a measurement grid on the bottom.
    • 1-Hand Held Magnifier
    • 1-Safety Test Tube (collection tube)
    • 3-Petri Dishes, 35mmD
    • 1-Forcep
    • 1-Pencil
    • 1-Compass
    • 1-Journal to record your findings.
    • 1-Backpack
  • Size: 16 x 12 x 8 in.
  • Ages 7+

Specifications

Grade 1
Product Type Supertek™ Field Explorer Kit
Includes 20X Stereo Microscope, Large Insect Net, Aqua Net, Bug Viewer, Hand Held Magnifier, Safety Test Tube, 3-Petri Dishes, Forcep, Pencil, Compass, Journal, Backpack
For Use With (Application) Field Trip Observations
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.