Learn More
Perkin Elmer US LLC PEEK One-Piece Fingertight Fitting
Catalog No. 502016450
Encompass_Preferred
The fittings are compatible with 1/16 O.D. tubing and have 10-32 threads. The fittings are biocompatible and sold in packages of 10.
Catalog No. 50-201-6450 Supplier Perkin Elmer US LLC Supplier No. ED020005
The fittings are compatible with 1/16 O.D. tubing and have 10-32 threads. The fittings are biocompatible and sold in packages of 10.