Learn More
Bachem Biotinyl-eAhx-Amyloid Beta Protein (1-42) Trifluoroacetate
SDP
Supplier:  Bachem 4090155.0500
Encompass_Preferred
Biotinyl-eAhx-Amyloid Beta Protein (1-42) Trifluoroacetate, Biotinyl-eAhx-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-Ile-Ala-OH, 1872440-40-8, C219H336N58O63S2, Mr 4853.56, 0.5mg. Biotinyl-eAhx-Ab42, Biotinyl-LC-Amyloid b-Protein (1-42). The flexible LC spacer increases the accessibility of the biotin moiety.
Catalog No. 50-194-6616