Learn More
Unchained Labs THE FUNDAMENTALS
SDP
Catalog No. NC3236569
Encompass
The Fundamentals - Kit to verify accuracy, precision, dynamic range and linearity. (500 μl verification standards, 5 glass vials)
Catalog No. NC3236569 Supplier Unchained Labs Supplier No. 7012029
The Fundamentals - Kit to verify accuracy, precision, dynamic range and linearity. (500 μl verification standards, 5 glass vials)