Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ The GLOBE Program Equipment: Hydrology Kits
SDPSCI_ED

Catalog No. S45030 Shop All LaMotte Company Products
Change view
Click to view available options
For Use With (Application):
Environmental education, drinking water, industrial water boiling and cooling, laundry and sanitation, water and wastewater
Food and beverage, environmental education, industrial water boiling and cooling, laundry and sanitation, water and wastewater
For Use With (Equipment):
Direct Reading Titrator
LRC Comparator
Includes:
Alkalinity Test Kit (50 BCG-MR indicator tablets, 60mL alkalinity titration reagent B, 5 to 10 to 15mL test tube with cap, direct reading titrator, 0 to 200 range, alkalinity endpoint color chart)
Nitrate Nitrogen Test Kit (120mL Mixed Acid reagent, 5g Nitrate reducing reagent, 60mL Phosphate Acid reagent, 5g Phosphate reducing reagent, 4 test tubes, 2 spoons, 1mL pipet, cap pull 'n' push, 5mL distilled water ampoule, axial reader, Nitrate-N and Phosphate Comparator)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) For Use With (Equipment) Includes
S45030 Environmental education, drinking water, industrial water boiling and cooling, laundry and sanitation, water and wastewater Direct Reading Titrator Alkalinity Test Kit (50 BCG-MR indicator tablets, 60mL alkalinity titration reagent B, 5 to 10 to 15mL test tube with cap, direct reading titrator, 0 to 200 range, alkalinity endpoint color chart)
S65412 Food and beverage, environmental education, industrial water boiling and cooling, laundry and sanitation, water and wastewater LRC Comparator Nitrate Nitrogen Test Kit (120mL Mixed Acid reagent, 5g Nitrate reducing reagent, 60mL Phosphate Acid reagent, 5g Phosphate reducing reagent, 4 test tubes, 2 spoons, 1mL pipet, cap pull 'n' push, 5mL distilled water ampoule, axial reader, Nitrate-N and Phosphate Comparator)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S45030 Supplier LaMotte™ Supplier No. 4491DR01
Add to Cart
Add to Cart

Test kits teach the real-world applications of hydrology

GLOBE is a worldwide hands-on, primary and secondary school-based science and education program. The equipment specified here is based on descriptions and specifications contained in The GLOBE Program Teacher's Guide. Skill level recommendations are indicated as Elementary, Middle, and High for each piece of specified equipment.

  • Water Alkalinity Kit
    • For use with environmental education, drinking water, industrial water boiling and cooling, laundry and sanitation, water and wastewater
    • Includes 50 BCG-MR indicator tablets, 60 mL alkalinity titration reagent B, 5 to 10 to 15 mL test tube with cap, direct reading titrator, 0 to 200 range and alkalinity endpoint color chart<
  • Water Nitrate Kit
    • For use with food and beverage, environmental education, industrial water boiling and cooling, laundry and sanitation, water and wastewater
    • Includes 120 mL mixed acid reagent, 5 g nitrate reducing reagent, 60 mL phosphate acid reagent, 5 g phosphate reducing reagent, 4 test tubes, 2 spoons, 1 mL pipet, cap, 5 mL distilled water ampoule, axial reader, nitrate-N and phosphate comparator

GLOBE, The GLOBE Program, and the GLOBE logo are the trademarks of the U.S. Government. Use of these trademarks and certification by this company that its products conform to the specificationd of The GLOBE Program does not imply endorsement of this company or its products by the U.S. Government.

Specifications

Grade 3+
For Use With (Equipment) Direct Reading Titrator
Product Type Kit, GLOBE Program Hydrology
Includes Alkalinity Test Kit (50 BCG-MR indicator tablets, 60mL alkalinity titration reagent B, 5 to 10 to 15mL test tube with cap, direct reading titrator, 0 to 200 range, alkalinity endpoint color chart)
For Use With (Application) Environmental education, drinking water, industrial water boiling and cooling, laundry and sanitation, water and wastewater
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.