Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ The GLOBE Program Equipment: Replacement Reagents for NPK Kit
SDPSCI_ED

Catalog No. S72347 Shop All LaMotte Company Products
Change view
Click to view available options
For Use With (Application):
Environmental education
To determine the reagent shelf life
Product Type:
GLOBE Program Replacement Reagents, Nitrate Wide Range Tablets
GLOBE Program Replacement Reagents, Phosphorous Tablets
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Product Type
S72347 To determine the reagent shelf life GLOBE Program Replacement Reagents, Nitrate Wide Range Tablets
S72346 Environmental education GLOBE Program Replacement Reagents, Phosphorous Tablets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S72347 Supplier LaMotte™ Supplier No. 3703AH
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Additional reagents for NPK Kit

  • GLOBE is a worldwide hands-on, primary and secondary school-based science and education program. The equipment specified here is based on descriptions and specifications contained in The GLOBE Program Teacher's Guide.

Specifications

Grade 3+
For Use With (Equipment) Soil NPK kit
Product Type GLOBE Program Replacement Reagents, Nitrate Wide Range Tablets
For Use With (Application) To determine the reagent shelf life
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.