Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Soil NPK Test Kit
SDPSCI_ED

Catalog No. S65951 Shop All LaMotte Company Products
Change view
Click to view available options
For Use With (Application):
Soil, environmental education
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
S65951 Soil, environmental education
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S65951 Supplier LaMotte™ Supplier No. 58806
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Unit dose tablet test for soil macronutrients

  • Simple diagramed instructions and laminated color chart
  • 50 tests each for nitrogen, phosphorus and potasium in soil
  • Originally developed for the GLOBE program
  • Test results for available nutrients listed as Low, Medium and High with approximations for lb/acre.

GLOBE, The GLOBE Program, and the GLOBE logo are the trademarks of the U.S. Government. Use of these trademarks and certification by this company that its products conform to the specificationd of The GLOBE Program does not imply endorsement of this company or its products by the U.S. Government.

Specifications

Grade 3+
Product Type Kit, NPK Soil Test
Includes The necessary apparatus, simple diagrammed instructions, and a laminated color chart
For Use With (Application) Soil, environmental education
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.