Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

The GLOBE Program™ Equipment
SCI_ED

Catalog No. S41580B
Change view
Click to view available options
Grade:
K to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S41580B K to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S41580B Supplier Laboratory Products Group (Lpg) Supplier No. 8TC110S

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

GLOBE is a worldwide hands-on, primary and secondary school-based science and education program. The equipment specified here is based on descriptions and specifications contained in The GLOBE Program Teacher's Guide. Skill level recommendations are indicated as Elementary, Middle, and High for each piece of specified equipment.

GLOBE, The GLOBE Program, and the GLOBE logo are the trademarks of the U.S. Government. Use of these trademarks and certification by this company that its products conform to the specificationd of The GLOBE Program does not imply endorsement of this company or its products by the U.S. Government.

TRUSTED_SUSTAINABILITY

Specifications

Grade Level Elementary, Intermediate - Middle School, High School
Grade K to 12
Product Type Calibration Thermometer
For Use With (Application) Study of atmosphere/climate for primary and secondary school based science and education program
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.