Learn More
Enterprise Technology Solutions The Gorilla Glue Company Epoxy, Gorilla Glue, w/Syringe, 0.85oz, Clear
SDP
Catalog No. 5000827324
Encompass_Preferred
Gorilla takes epoxy to the Gorilla tough level. With superior solvent and water-resistance, this long-lasting adhesive is incredibly strong and durable for household and automotive repairs alike. Easy-to-use syringe keeps the epoxy resin and hardener separate so it is easy to dispense. Two-part, gap-filling formula easily bonds steel, aluminum, glass, wood, ceramic, tile and most plastics. The 5-minute set time gives you plenty of repositioning time for the perfect fit. Gorilla Epoxy dries in an ideal, clear finish that makes projects look spotless.
Catalog No. 50-008-27324 Supplier Enterprise Technology Solutions Supplier No. GOR4200101
Gorilla takes epoxy to the Gorilla tough level. With superior solvent and water-resistance, this long-lasting adhesive is incredibly strong and durable for household and automotive repairs alike. Easy-to-use syringe keeps the epoxy resin and hardener separate so it is easy to dispense. Two-part, gap-filling formula easily bonds steel, aluminum, glass, wood, ceramic, tile and most plastics. The 5-minute set time gives you plenty of repositioning time for the perfect fit. Gorilla Epoxy dries in an ideal, clear finish that makes projects look spotless.