Learn More
Enterprise Technology Solutions The Gorilla Glue Company Glue Gun,Full,Dual Temp,12-Watt,w/Rubber Handle,OE/BK
SDP
Catalog No. 5000826429
Encompass_Preferred
Dual Temp Full-Size Glue Gun is an ideal tool for crafts and repairs requiring more glue output and volume. Lightweight design, comfort grip and built-in stand make it easy to use. Easy-squeeze trigger and long, insulated nozzle allow a precise application on whatever project you are working on. Press switch to high temperature for wood, plastic, metal, glass and more. Flip to low temperature for floral, fabric, foam, jewelry and more. Glue gun uses full-size, 0.43" glue sticks (not included). Safety features include a safety fuse and cord protection to prevent cord from shorting out.
Catalog No. 50-008-26429 Supplier Enterprise Technology Solutions Supplier No. GOR100424
Dual Temp Full-Size Glue Gun is an ideal tool for crafts and repairs requiring more glue output and volume. Lightweight design, comfort grip and built-in stand make it easy to use. Easy-squeeze trigger and long, insulated nozzle allow a precise application on whatever project you are working on. Press switch to high temperature for wood, plastic, metal, glass and more. Flip to low temperature for floral, fabric, foam, jewelry and more. Glue gun uses full-size, 0.43" glue sticks (not included). Safety features include a safety fuse and cord protection to prevent cord from shorting out.