Learn More
Enterprise Technology Solutions The Gorilla Glue Company Glue, Original, Indoor/Outdoor, 2 oz, Clear
SDP
Catalog No. 5000826232
Encompass_Preferred
Original Formula Gorilla Glue is tough, expanding and sticks to everything. It is incredibly strong and ideal for indoor and outdoor applications. Known for its industrial holding power and versatility, regardless of the surface, Gorilla Glue is 100 percent waterproof and doesn't break down when exposed to outdoor elements. It easily bonds wood, stone, metal, ceramic, foam, glass, concrete and much more. The water-activated polyurethane formula expands three times into the materials to form an incredibly strong bond. Bond holds through hot and cold temperatures. Clamp time is 1 to 2 hours.
Catalog No. 50-008-26232 Supplier Enterprise Technology Solutions Supplier No. GOR5000201
Original Formula Gorilla Glue is tough, expanding and sticks to everything. It is incredibly strong and ideal for indoor and outdoor applications. Known for its industrial holding power and versatility, regardless of the surface, Gorilla Glue is 100 percent waterproof and doesn't break down when exposed to outdoor elements. It easily bonds wood, stone, metal, ceramic, foam, glass, concrete and much more. The water-activated polyurethane formula expands three times into the materials to form an incredibly strong bond. Bond holds through hot and cold temperatures. Clamp time is 1 to 2 hours.