Learn More
Enterprise Technology Solutions The Gorilla Glue Company Super Glue,Gel,Thick,Quick-Dry,Tube,3g,4/PK,Clear
SDP
Catalog No. 5000826426
Encompass_Preferred
Gorilla Super Glue Gel is thicker and more controlled, developed for long-lasting repairs in an instant. Quick-dry formula dries within 10-45 seconds. This super glue works on a variety of materials, including plastic, metal, stone, ceramic, paper, rubber and more. Super glue gel is not recommended for use on polypropylene plastic or similar materials.
Catalog No. 50-008-26426 Supplier Enterprise Technology Solutions Supplier No. GOR105800
Gorilla Super Glue Gel is thicker and more controlled, developed for long-lasting repairs in an instant. Quick-dry formula dries within 10-45 seconds. This super glue works on a variety of materials, including plastic, metal, stone, ceramic, paper, rubber and more. Super glue gel is not recommended for use on polypropylene plastic or similar materials.