Learn More
Enterprise Technology Solutions The Gorilla Glue Company Super Glue,Original,Waterproof,Indoor&Outdoor,Poly,4oz,Clear
SDP
Catalog No. 5000825721
Encompass_Preferred
Original Gorilla Glue features incredible strength and unmatched versatility. Polyurethane formula expands up to three times its original size when activated by water for a permanent bond. Waterproof design is ideal for both indoor and outdoor use. Industrial-strength adhesive works effectively in extreme temperatures ranging from -40 degrees to 200 degrees Fahrenheit.
Catalog No. 50-008-25721 Supplier Enterprise Technology Solutions Supplier No. GOR5000408
Original Gorilla Glue features incredible strength and unmatched versatility. Polyurethane formula expands up to three times its original size when activated by water for a permanent bond. Waterproof design is ideal for both indoor and outdoor use. Industrial-strength adhesive works effectively in extreme temperatures ranging from -40 degrees to 200 degrees Fahrenheit.