Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ The Great Mum Mix Up
SDPSCI_ED

Catalog No. S001096 Shop All EDVOTEK Products
Change view
Click to view available options
Includes:
FlashBlue™ DNA Stain
SYBR Safe DNA Stain
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S001096 SYBR Safe DNA Stain
S001095 FlashBlue™ DNA Stain
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S001096 Supplier EDVOTEK™ Supplier No. 125S
Add to Cart
Add to Cart

Students will use agarose gel electrophoresis to differentiate between genetically engineered true blue chrysanthemums and artificially-dyed flowers

  • Includes FlashBlue™ and SYBR™ Safe DNA Stain.
  • Hands-on use of core biotech tools to detect and identify genetically modified plants in simulated samples.
  • Understand how electrophoresis separates differently-sized molecules.
  • Perform agarose gel electrophoresis.
  • Meets NGSS HS DCI LS1.A, LS3.A, LS3.B, and ETS1.B through the discussion of the structure and function of DNA, heredity and variance of traits, and use of biotechnology to make genetic modifications.
  • Group Size: For 8 Gels.
  • Kit Includes: Instructions, Ready-to-Load QuickStrip™ DNA Samples, UltraSpec-Agarose™, Electrophoresis Buffer (50X), Practice Gel Loading Solution, FlashBlue™ DNA Stain.
  • All You Need: DNA Electrophoresis, Micropipettes: 5-50 μL (Optional), Blue Light Transilluminator, and Microwave or Hot Plate.
  • Storage: Room Temperature Stable. Storage of Ready-to-Load QuickStrip™ samples in the Refrigerator is Recommended.

Specifications

Grade Level Intermediate - Middle School, High School
Grade 9+
Includes SYBR Safe DNA Stain
Class Size 8 Lab Groups
Product Type Electrophoresis Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.