Learn More
Anton Paar The high-precision thermometers from Anton Paar are designed for the most accurate temperature measurements, comparison calibrations and fixed point calibrations in industry, laboratories and research. Combined with calibrated pla
Catalog No. 5000580496
Encompass_Preferred
Measuring range
Temperature: −260 °C to +962 °C (−436 °F to +1764 °F)
Resistance: 0 Ω to 440 Ω
Resolution
Temperature: 0.1 mK (Pt 100)
Resistance: 40 μΩ
Measurement uncertainty
Temperature: < 1 mK (Pt 100)
Resistance: < 400 μΩ
Catalog No. 50-005-80496 Supplier Anton Paar Supplier No. 26878
Measuring range
Temperature: −260 °C to +962 °C (−436 °F to +1764 °F)
Resistance: 0 Ω to 440 Ω
Resolution
Temperature: 0.1 mK (Pt 100)
Resistance: 40 μΩ
Measurement uncertainty
Temperature: < 1 mK (Pt 100)
Resistance: < 400 μΩ