Learn More
Perkin Elmer US LLC Communication Cable for the 50 mm Integrating Spheres
Catalog No. 502016331
Encompass_Preferred
The high sensitivity 50 mm Transmission/Reflectance Sphere is compatible with LAMBDA 365 spectrometers. It is used for measuring diffuse reflectance (specular component included SCI), total transmittance or hemispherical transmittance, and performing color analysis. It has a spectral range of 230 to 1100 nm (UV and visible). It has a port size of 10 mm. The recommended spectral bandwidth setting is 5 nm. Recommended for solid color analysis.
Catalog No. 50-201-6331 Supplier Perkin Elmer US LLC Supplier No. N4101501
The high sensitivity 50 mm Transmission/Reflectance Sphere is compatible with LAMBDA 365 spectrometers. It is used for measuring diffuse reflectance (specular component included SCI), total transmittance or hemispherical transmittance, and performing color analysis. It has a spectral range of 230 to 1100 nm (UV and visible). It has a port size of 10 mm. The recommended spectral bandwidth setting is 5 nm. Recommended for solid color analysis.