Learn More
Bachem Fmoc-Asp-ODmb
SDP
Supplier:  Bachem 4018284.0001
Encompass_Preferred
Fmoc-Asp-ODmb, 155866-25-4, C28H27NO8, Mr 505.52, 1g. The highly acid-labile Dmb ester can be cleaved, e.g., with 1%TFA/DCM.
Catalog No. 50-194-7120