Learn More
Bachem Penetratin Trifluoroacetate
SDP
Supplier:  Bachem 4036091.0001
Encompass_Preferred
Penetratin Trifluoroacetate, H-Arg-Gln-Ile-Lys-Ile-Trp-Phe-Gln-Asn-Arg-Arg-Met-Lys-Trp-Lys-Lys-NH2, 214556-79-3, C104H169N35O19S, Mr 2245.78, 1mg. Antennapedia Homeobox (43-58) amide, Antennapedia Peptide. The highly efficient cell-penetrating peptide penetratin derived from the third helix of the Antennapedia protein homeodomain also showed antimicrobial and antifungal activity.
Catalog No. 50-194-6678