Learn More
Enterprise Technology Solutions The HON Company Storage Cabinet, Metal, Tray Kits, 30"x18"x52-1/2", Loft
SDP
Catalog No. 5000812115
Encompass_Preferred
To minimize clutter and maximize efficiency, Flagship Modular Storage Cabinet supports active to archival to communal storage that will organize everything but your thoughts. Multipurpose filing solution with storage bins offers versatile ways to maximize space, define boundaries and bring people closer to the things they need to stay productive. All-metal storage cabinet is pre-configured with six 3" bins, six 6" bins, two 12" bins and rails (all included). Additional Tray Kits (3", 6" or 12" high bins) are sold separately. Trays and rails can be easily rearranged for greater efficiency. Doors are equipped with HON "One Key" interchangeable core-removable locks, full radius handles, vertical stiffener and positive door stops. Top and base are reinforced for durability. Cabinet features four adjustable leveling glides for stability. It also meets or exceeds ANSI/BIFMA and ISTA standards and is certified SCS Indoor Advantage Gold.
Catalog No. 50-008-12115 Supplier Enterprise Technology Solutions Supplier No. HONSC185230LGLO
To minimize clutter and maximize efficiency, Flagship Modular Storage Cabinet supports active to archival to communal storage that will organize everything but your thoughts. Multipurpose filing solution with storage bins offers versatile ways to maximize space, define boundaries and bring people closer to the things they need to stay productive. All-metal storage cabinet is pre-configured with six 3" bins, six 6" bins, two 12" bins and rails (all included). Additional Tray Kits (3", 6" or 12" high bins) are sold separately. Trays and rails can be easily rearranged for greater efficiency. Doors are equipped with HON "One Key" interchangeable core-removable locks, full radius handles, vertical stiffener and positive door stops. Top and base are reinforced for durability. Cabinet features four adjustable leveling glides for stability. It also meets or exceeds ANSI/BIFMA and ISTA standards and is certified SCS Indoor Advantage Gold.