Learn More
Key Scientific Products THE HOOK / EACH, Angled for Easier Capture of Cryo Bead
SDP
Catalog No. NC0861795
Encompass_Preferred
"The Hook" has a special angle to assist in easier capture of a single CryoCare bead or other freeze systems for recovery of stored frozen microorganisms. It can also be used in a Mycology Lab as a teasing needle.
Catalog No. NC0861795 Supplier Key Scientific Products Supplier No. KSC4010
"The Hook" has a special angle to assist in easier capture of a single CryoCare bead or other freeze systems for recovery of stored frozen microorganisms. It can also be used in a Mycology Lab as a teasing needle.