Learn More
Taylor Wharton 16" Cryogenic Roller Base
Catalog No. NC1609001
Encompass_Preferred
For easier mobility of dewars 16" in diameter or less.
Catalog No. NC1609001 Supplier Taylor Wharton Supplier No. R0188C00
For easier mobility of dewars 16" in diameter or less.