Learn More
Electron Microscopy Sciences The mBox
SDP
Catalog No. 50949191
Encompass_Preferred
*mBox Only* mBox in Microarray Slide Handling. Measures 5" L x 3½"W x 1½" H (127mm x 89mm x 38mm) Immerse 20 slides in liquid during blocking, denaturating, and post-arraying. It separates and prevents slide damage during shipping and processing. Optimally designed rack spacing for easy slide removal.
Catalog No. 50-949-191 Supplier Electron Microscopy Sciences Supplier No. 7218801
*mBox Only* mBox in Microarray Slide Handling. Measures 5" L x 3½"W x 1½" H (127mm x 89mm x 38mm) Immerse 20 slides in liquid during blocking, denaturating, and post-arraying. It separates and prevents slide damage during shipping and processing. Optimally designed rack spacing for easy slide removal.