Learn More
Perkin Elmer US LLC Stainless Steel Test Sample
Catalog No. 502016341
Encompass_Preferred
The Measuring System requires the following calibration test samples to ensure optimal performance: PMMA, Stainless Steel, PBC, HDPE, Polycarb, PET Film, and G10 Epoxy. The test samples come standard with this DMA 8000 system, but they can also be ordered individually as needed.
Catalog No. 50-201-6341 Supplier Perkin Elmer US LLC Supplier No. N5337016
The Measuring System requires the following calibration test samples to ensure optimal performance: PMMA, Stainless Steel, PBC, HDPE, Polycarb, PET Film, and G10 Epoxy. The test samples come standard with this DMA 8000 system, but they can also be ordered individually as needed.