Learn More
Martor Secupro Megasafe
Catalog No. NC9935939
Encompass_Preferred
The Megasafe is an ambidextrous safety cutter with a fully automatic retractable blade. It features almost an inch of blade extension and a durable metal body to make it the most versatile safety knife on the market.-Fully automatic retractable blade for safety-Ergonomically friendly ambidextrous slider mechanism to engage the blade-Durable body made of aluminum-Safer alternative to fixed blade utility knives for general cutting tasks-Just under 1" of blade extension
Catalog No. NC9935939 Supplier Martor Supplier No. 116006.02
The Megasafe is an ambidextrous safety cutter with a fully automatic retractable blade. It features almost an inch of blade extension and a durable metal body to make it the most versatile safety knife on the market.-Fully automatic retractable blade for safety-Ergonomically friendly ambidextrous slider mechanism to engage the blade-Durable body made of aluminum-Safer alternative to fixed blade utility knives for general cutting tasks-Just under 1" of blade extension