Learn More
Bachem Fmoc-Orn(Dde)-OH
SDP
Supplier:  Bachem 4027859.0001
Encompass_Preferred
Fmoc-Orn(Dde)-OH, 269062-80-8, C30H34N2O6, Mr 518.61, 1g. The Nd-protecting group Dde is orthogonal to both Fmoc and Boc which allows on-resin modification of ornithine side chains during Fmoc/tBu-based SPPS. The applications of Dde side-chain protection include synthesis of branched or di-epitopic peptides, preparation of MAP core molecules and lipo-MAPS, the synthesis of cyclic peptides, TASP molecules, and templates for combinatorial chemistry and synthetic proteins. Dde can be cleaved by 2% hydrazine in DMF.
Catalog No. 50-194-7245