Learn More
Perkin Elmer US LLC Provectus Balance Controller Unit
Supplier:  Perkin Elmer US LLC N5340362
Encompass_Preferred
The new Provectus 6500Autobalance is a microprocessor-controlled ultra-microbalance capable of weighing samples from 0.1 µg to 1000 mg. Provectus offers a wide range of features, including high performance, fully automated operation, automatic calibration, and user convenience. Used in production areas for the calibration of precision micropipettes, weighing precious metals, food stuffs, dispensing modern drugs, and preparing costly instrument standards in the environmental and biotechnology areas where accurate and precise measurements are vital to their results. It features an easy-to-use keyboard with a graphical OLED display and microcomputer control. Weighing changer has right-or left-hand access and is separate from microcomputer controller for maximum flexibility in any laboratory environment. Pan arrests are standard. Built-in microprocessor functions include Automatic Tare, Automatic Calibration, and Automatic Range Selection. Additional features include storage capacity for
Catalog No. 50-201-6537
Add to Cart
Request a Quote