Learn More
Perkin Elmer US LLC Near Infrared Reflectance Accessory (NIRA)
Supplier:  Perkin Elmer US LLC L1240050
Encompass_Preferred
The NIRA is an intelligent sampling system designed for reflectance measurements of solids and transflectance measurements of liquids. The technique is non-destructive and particularly useful for undiluted powders and granular materials. The accessory can be used for a wide variety of sample types in various containers and includes self-referencing for increased reproducibility and ease of use. Features of the NIRA include: Integrated, zero alignment, zero set-up design. Once locked into the sample compartment the NIRA accessory becomes an integral part of the system and is ready to use. Horizontal sample platform for convenient sample positioning. Integrated, high performance stabilized InGaAs detector system standard. Range of optional accessories such as the sample spinner are available for inhomogeneous solids or liquids samples. Removed and replaced in seconds via quick-release lever with automatic parameter setup. The NIRA is supplied with a standard accessories kit including a
Catalog No. 50-201-6518
Add to Cart
Request a Quote