Learn More
Perkin Elmer US LLC Near Infrared Reflectance Accessory (NIRA) for Frontier, Sample Compartment
Supplier:  Perkin Elmer US LLC L1280313
Encompass_Preferred
The Near Infrared Reflectance Accessory (NIRA) is designed for diffuse reflectance measurements of light-scattering solids, liquids, and powders using the Frontier FT-NIR or FT-IR/NIR spectrometer.
Catalog No. 50-201-6503
Add to Cart
Request a Quote