Learn More
SOMNI Scientific™ The Non-Rebreathing Dental Table Support System
SDP
Catalog No. 04777176 Shop All Somni Scientific Products
Change view
Click to view available options
For Use With (Application):
Research Animal Inhalant Anesthesia
Catalog No. For Use With (Application)
04-777-176 Research Animal Inhalant Anesthesia
1 options
Catalog No. 04-777-176 Supplier SOMNI Scientific™ Supplier No. PH4020
Add to Cart
Add to Cart
Critical accessory used with non-rebreathing circuits during dentistry. It provides stability and rotation without kinking the circuit or bag and anchors directly to the dental grate.
  • Critical accessory used with non-rebreathing circuits during dentistry
  • Provides stability and rotation
  • Will not kink the circuit or bag
  • Anchors directly to the dental grate
  • Easy installation
Specifications
For Use With (Application) Research Animal Inhalant Anesthesia
For Use With (Equipment) Non-Rebreathing Circuits During Dentistry