Learn More
Perkin Elmer US LLC Knurled Nut for 2.4 mm O.D. Tubing
Supplier:  Perkin Elmer US LLC B3001335
Encompass_Preferred
The nut is part of an assembly that requires front ferrule (B3001333), and back ferrule (B3001334). These components are compatible with tubing B0017998 which is sold by the meter (m).
Catalog No. 50-201-6307