Learn More
Bachem Sex Pheromone Inhibitor iPD1 Trifluoroacetate
SDP
Catalog No. 501947520
Encompass_Preferred
Sex Pheromone Inhibitor iPD1 Trifluoroacetate, H-Ala-Leu-Ile-Leu-Thr-Leu-Val-Ser-OH, 120116-56-5, C39H72N8O11, Mr 829.05, 5mg. The octapeptide ALILTLVS is produced by Streptococcus faecalis donor strains harboring bacteriocin plasmid pPD1. It inhibits the activity of sex pheromone cPD1. The non-polar peptide iPD1 is also used for calibration in mass spectrometry.
Catalog No. 50-194-7520 Supplier Bachem Supplier No. 4018007.0005
Add to Cart
Add to Cart
Sex Pheromone Inhibitor iPD1 Trifluoroacetate, H-Ala-Leu-Ile-Leu-Thr-Leu-Val-Ser-OH, 120116-56-5, C39H72N8O11, Mr 829.05, 5mg. The octapeptide ALILTLVS is produced by Streptococcus faecalis donor strains harboring bacteriocin plasmid pPD1. It inhibits the activity of sex pheromone cPD1. The non-polar peptide iPD1 is also used for calibration in mass spectrometry.