Learn More
Bachem Fmoc-D-Asp(OMpe)-OH
SDP
Supplier:  Bachem 4039622.0005
Encompass_Preferred
Fmoc-D-Asp(OMpe)-OH, 1926162-97-1, C25H29NO6, Mr 439.51, 5g. The OMpe-side chain protecting group significantly reduces aspartimide formation, an undesired side-chain reaction encountered with the repetitive piperidine treatment needed for Fmoc removal.
Catalog No. 50-194-7125