Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mechanix Wear™ The Original™ Red Mechanics Gloves

Catalog No. 19030306C Shop All Mechanix Wear Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-030-306C Large
19-030-306B Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-030-306C Supplier Mechanix Wear™ Supplier No. MG02010
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Original™ gloves are built with carbon-infused synthetic leather for touchscreen capabilities, and breathable TrekDry™ to deliver a blend of dexterity and durability.

  • TrekDry™ is form-fitting to help keep hands cool and comfortable
  • Adjustable thermoplastic rubber (TPR) wrist closures create a secure fit
  • Reinforced thumb and index finger improve durability
  • Touchscreen capable synthetic leather for a combination of dexterity and durability
  • Ideal for maintenance repairs, equipement operation, and DIY home improvement
  • Machine washable

Specifications

Back Material TrekDry™
Cuff Material Thermoplastic Rubber (TPR)
Coating Material Uncoated
Color Red
Hand Specific
Material Synthetic Leather
Product Type Mechanic's Gloves
Size Large
For Use With (Application) Automotive Work, Maintenance Repairs, Equipment Operation, and DIY Home Improvements
Glove Cuff Style Hook and Loop
Series The Original™
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.