Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leica Microsystems The pE-300lite SB Direct Fit

Catalog No. 10376201 Shop All Leica Microsystems Products
Change view
Click to view available options
Type:
Illuminator
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
10-376-201 Illuminator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-376-201 Supplier Leica Microsystems Supplier No. 11504267
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The pE-300lite SB Direct Fit LED fluorescence light source is ideal for stereomicroscopes.

  • For use with direct coupling
  • Includes light source, control pod, power supply
  • The pE 300 lite SB light source is ideally suited for regular routine fluorescence applications offering intense, broad-spectrum LED illumination
  • Spectral coverage (365-650nm) is from the UV (DAPI excitation) to the red region (Cy5 excitation)
  • The system is a mercury-free alternative that is safer, more controllable and precise than traditional bulb illumination options
  • Operation is by a remote manual control pod with instant on/off and control of irradiance from 0-100%
TRUSTED_SUSTAINABILITY

Specifications

Type Illuminator
Autoclavable Not Autoclavable
Bulb Type LED
Dimensions (L x W x H) 186 x 77 x 162 mm
Excitation/Emission Broad spectrum (365-550 nm)
Voltage 100/240 VAC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.