Learn More
Bachem Gastric Juice Peptide Fragment Trifluoroacetate
SDP
Supplier:  Bachem 4030660.1000
Encompass_Preferred
Gastric Juice Peptide Fragment Trifluoroacetate, H-Gly-Glu-Pro-Pro-Pro-Gly-Lys-Pro-Ala-Asp-Asp-Ala-Gly-Leu-Val-OH, 137525-51-0, C62H98N16O22, Mr 1419.56, 1mg. BPC 157. The pentadecapeptide BPC-157 (GEPPPGKPADDAGLV) shows a protective effect on stomach and duodenum when administered to stress ulcers, cysteamine-duodenal ulcers and ethanol lesions. Application of BPC 157 improved tendon healing.
Catalog No. 50-194-6927