Learn More
Bachem (Glu17·21·24)-Osteocalcin (1-49) (human) Trifluoroacetate
SDP
Supplier:  Bachem 4063516.0100
Encompass_Preferred
(Glu17·21·24)-Osteocalcin (1-49) (human) Trifluoroacetate, H-Tyr-Leu-Tyr-Gln-Trp-Leu-Gly-Ala-Pro-Val-Pro-Tyr-Pro-Asp-Pro-Leu-Glu-Pro-Arg-Arg-Glu-Val-Cys-Glu-Leu-Asn-Pro-Asp-Cys-Asp-Glu-Leu-Ala-Asp-His-Ile-Gly-Phe-Gln-Glu-Ala-Tyr-Arg-Arg-Phe-Tyr-Gly-Pro-Val-OH, 1927927-11-4, C266H381N67O76S2, Mr 5797.49, 0.1mg. Osteocalcin (1-49) (human) (decarboxylated). The peptide hormone osteocalcin is involved not only in bone formation, it also plays an important role in glucose metabolism and could regulate testosteron. In mice, only the completely decarboxylated form of the peptide shows the latter hormonal activities, whereas in humans, osteocalcin, partially decarboxylated osteocalcins, and the uncarboxylated peptide seem to be involved. Generally, obese individuals have been shown to have lower osteocalcin(s) levels than non-obese controls, and type 2 diabetic individuals have lower plasma osteocalcin than non-diabetic individuals.
Catalog No. 50-194-6911