Learn More
Bachem H-Arg-Val-Leu-psi(CH2NH)Phe-Glu-Ala-Nle-NH2
SDP
Supplier:  Bachem 4030746.0001
Encompass_Preferred
H-Arg-Val-Leu-psi(CH2NH)Phe-Glu-Ala-Nle-NH2, 167875-35-6, C40H69N11O8, Mr 832.06, 1mg. The peptide RVL-psi[CH2NH]FEA-Nle-amide containing a reduced peptide bond was found to be a competitive inhibitor of the HIV-1 protease (Ki = 50 nM).
Catalog No. 50-194-6950