Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

The Perfect Plan Book
SDPSCI_ED

Catalog No. S95199
Change view
Click to view available options
Grade:
K to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S95199 K to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S95199 Supplier Carson Dellosa Publishing Supplier No. 0742400271

missing translation for 'validMainframeMsg'

Revised and Improved!

This fantastic planner features 88 planning pages with blank headings so you can fill in your schedule, rules for writing, spaces to list absences, a reminder area, and more.

  • Flexible format allows you to record all the week's information for quick and easy access
  • You'll also find student information pages, a year-at-a-glance page, birthday pages, seating charts, and more
  • Features adaptable spaces, ruled lines, spiral-bound convenience, and space for at least eight subjects

Order Info

  • Grade level: K-6
  • Soft cover, 88 pages

Specifications

Grade K to 12
Format Book
Product Type Record/Plan Book
For Use With (Application) Features adaptable spaces, ruled lines, spiral-bound convenience
Subject General Education
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.